Learn More
Enzo Life Sciences CYTO-ID Autophagy detection kit 2.0 (50 tests)
SDP
Supplier:  Enzo Life Sciences ENZKIT1750050
Encompass_Preferred
CYTO-ID Autophagy Detection Kit 2.0 measures autophagic vacuoles and monitors autophagic flux in lysosomally inhibited live cells using a novel dye that selectively labels accumulated autophagic vacuoles. The dye has been optimized through the identification of titratable functional moieties that allow for minimal staining of lysosomes while exhibiting bright fluorescence upon incorporation into pre-autophagosomes, autophagosomes, and autolysosomes (autophagolysosomes). The assay offers a rapid and quantitative approach to monitoring autophagy in live cells without the need for cell transfection. Mechanism of Action The probe is a cationic amphiphilic tracer (CAT) dye that rapidly partitions into cells in a similar manner as drugs that induce phospholipidosis. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection, HTS .
Catalog No. 50-201-3856