Learn More
THERMO SCI LSG 3P ATTUNE ATTUNE NXT AFC BG-LASER EACH
Supplier: THERMO SCI LSG 3P ATTUNE A28995
The Attune NxT Flow Cytometer has expanded functionalities to drive your research forward. Designed and developed to remove common barriers associated with flow cytometry, the evolutionary capabilities of the Attune NxT Flow Cytometer offer adaptable optical configuration options so you can get the most out of your multicolor analysis.• New CytKick and CytKick Max autosampler—more efficiency for high-throughput assays• New 21 CFR part 11 compliant software for regulated laboratories on Windows 10 operating system• Improved workflow for run protocol and instrument settings managementFast flowThe innovative combination of acoustic focusing technology coupled with traditional hydrodynamic focusing results in acquisition speeds up to 10X faster than other flow cytometers. This technology reduces the time needed to obtain results.• True 35,000 event/sec (+10% coincidence rates)• Maximum electronic speed of 65,000 events/sec• Efficiently run traditionally long assays