Learn More
Johannes Lieder Arrangement and Types of Vascular Bundles (steles)
6250 Arrangement and Types of Vascular Bundles (steles), 13 microscope slides
Supplier: Johannes Lieder 6250
Description
- 6250 Arrangement and Types of Vascular Bundles
- 13 Microscope Slides
- Protostele. Psilotum, stem t.s.
- Actinostele. Lycopodium, stem t.s.
- Polystele. Pteridium, rhizome t.s. showing concentric vascular bundles with inner xylem
- Ectophloic siphonostele. Osmunda, rhizome t.s.
- Amphiphloic siphonostele. Adiantum, rhizome t.s.
- Dictyostele. Polypodium, rhizome t.s.
- Eustele. Ranunculus, stem t.s., open collateral bundles
- Eustele. Lamium, stem t.s.
- Eustele. Cucurbita pepo, stem t.s., bicollateral bundles
- Atactostele. Zea mays, stem t.s., closed collateral bundles
- Arrangement of bundles similar to atactostele in a dicot plant. Podophyllum, stem t.s.
- Concentric vascular bundles with outer xylem. Convallaria, rhizome t.s.
- Radial concentric vascular bundle. Ranunculus, root t.s.
Specifications
Botany | |
Cross and Longitudinal Section | |
Prepared Slide |
Elastin Stained Glass | |
Microscope Slides |