Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Arium™ Mini Essential for Production of ASTM Type 1 Ultrapure Water with UV

Compact Arium™ mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 liters per day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.

Supplier:  Sartorius H2OMUUVTUS

Request Bulk Order

Catalog No. 14-558-730


Add to Cart

Description

Description

  • Reliable: Delivers consistently high water quality for reliable and reproducible results
  • Intuitive: Touch-activated color display with direct access to all important dispensing options
  • Innovative: saves time-intensive tank cleaning
  • Compact: Space-saving, with a width of only 28 cm, UV Lamp for Critical Analyses (185 | 254 nm)
  • Optimal temperature gradient due to horizontal installation
  • Degradation of total organic carbons (TOCs) by irradiation (< 5 ppb)
  • Inactivates microorganisms by damaging their DNA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EU Declaration of Conformity, CSA Declaration of Conformity, QA Certificate
10 L
Pretreated
Type 1
< 5 ppb
Lab Water System with UV Lamp
0 to 6.9 bar; (0 to approx. 100 psi); recommended > 2 bar (>29 psi)
20.8 x 11 x 20 in.
509 mm
100/240 V
< 100 μS/cm, compensated to 25°C
Water pretreated by reverse osmosis, distillation or deionization
Up to 1.0 L/min.
18.2 Mohm.cm at 25°C
<10 ppb
Touch activated
Arium mini accessories
280 mm
100/240 VAC 50/60 Hz 2.0 A max
Input supply voltage VDC +24 (±10%) Current consumption, max. A 3.0
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit