Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Aquasearcher™ pH Meter AB41PH
The Aquasearcher™ pH Meter AB41PH is an advanced, research-grade benchtop pH meter with electrode, offering accurate, repeatable results with selectable resolution and an intelligent i-Steward system.
$1314.00
Specifications
Accuracy (mV) | ±0.3 |
---|---|
Accuracy (pH) | ±0.002 |
Accuracy (Temperature) | ±0.3°C |
Data Management Details | Datalog with Time and Date Stamp |
Outputs | USB, RS-232 (PC), RS-232 (Printer) |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
S05294
|
OHAUS™
30589831/EMD |
AB41PH Meter, ST410 Electrode, STTEMP Probe, Electrode Holder, AS20 Magnetic Stirrer, 4 pH Buffer Mini Kits (50 mL) |
Each for $1,314.00
|
|
|||||
S05295
|
OHAUS™
30589830/EMD |
AB41PH Meter, Electrode Holder |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
Description
- Equipped with a large 6.5 in. LCD display and a touch keypad
- Features a 1000 item memory and password management for GLP documentation
- Provides connectivity and data output capabilities with RS232 and USB interface
- i-Steward ensures repeatable and consistent measurement accuracy
- Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
- Choose between just the AB41PH meter with electrode holder and a full kit including AB41PH meter, ST410 electrode, STTEMP probe, electrode holder, AS20 magnetic stirrer, and 4 pH buffer mini kits (50 mL)
Specifications
±0.3 | |
±0.3°C | |
USB, RS-232 (PC), RS-232 (Printer) | |
Offset | |
0.001, 0.01, 0.1 | |
Up to 5 | |
LCD, 6.5 in. Segment and Dot Matrix | |
1000 Sets |
±0.002 | |
Datalog with Time and Date Stamp | |
Auto-stop, Timed, Continuous | |
-2 to 20 | |
±2000 | |
-10°C to +125°C | |
LCD | |
1000 Measurement, 10 Calibration |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title