Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Aquasearcher™ pH Meter AB33PH
The Aquasearcher™ pH Meter AB33PH is a highly reliable and user-friendly pH benchtop meter with electrode (ST310) for standard laboratory applications. Designed to measure pH and oxidation-reduction potential with temperature measurements.
$758.00
Specifications
Accuracy (mV) | ±1 |
---|---|
Accuracy (pH) | ±0.01 |
Accuracy (Temperature) | ±0.5°C |
Data Management Details | Datalog with Time and Date Stamp |
Outputs | USB, RS-232 (PC), RS-232 (Printer) |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
S05296
|
OHAUS™
30589827/EMD |
AB33PH Meter, ST310 Electrode, Electrode Holder, 4 pH Buffer Mini Kits (50 mL) |
Each for $758.00
|
|
|||||
S05297
|
OHAUS™
30589826/EMD |
AB33PH Meter, Electrode Holder |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
Description
- Equipped with a large 6.5 in. LCD display and a touch keypad
- Features a 1000 item memory for pH measurements and calibration trail
- Provides connectivity and data output capabilities with RS232 and USB interface
- i-Steward monitors the condition of electrodes to ensure repeatable and consistent measurement accuracy
- Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
- Auto endpoint mode and auto buffer recognition makes calibration easy
- Choose between just the AB33PH meter with electrode holder and a full kit including AB33PH meter, ST310 electrode, electrode holder, 4 pH buffer mini kits (50 mL)
Specifications
±1 | |
±0.5°C | |
USB, RS-232 (PC), RS-232 (Printer) | |
Linear | |
0.01, 0.1 | |
Up to 3 | |
LCD, 6.5 in. Segment and Dot Matrix | |
1000 Sets |
±0.01 | |
Datalog with Time and Date Stamp | |
Auto-stop, Continuous | |
-2 to 16 | |
±2000 | |
-5°C to +110°C | |
LCD | |
1000 Measurement, Last Calibration |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title