Learn More
American Innotek Disposa-John™ Portable Restroom
Everything needed to safely dispose of solid human waste
Supplier:  American Innotek BR500
Catalog No. 19-805-951
Add to Cart
Includes: Toilet paper and antiseptic wipe.
Description
Description
  • Triple layer barrier bag and liner with easy-to-use design
  • Special blend of polymers and enzymes instantly begin to break down waste into a deodorized gel
  • Virtually odor free
  • Trash container safe; meets state, EPA, and provincial disposal requirements
  • Suitable for disposal of feminine hygiene products
Specifications
Specifications
Portable Restroom
Toilet paper and antiseptic wipe
Product Suggestions
Product Suggestions