Learn More
Anton Paar Alex 500 Alcohol and Extract Meter for Beer, Wine, Cider, Spirits, Liqueurs, and Sake
Supplier: Anton Paar 154512
Alex500 is a compact alcohol and extract meter that frees craft beverage producers from the need for external laboratories. Besides alcohol and extract content it determines related parameters, such as calories or degree of fermentation. Alex500 accurately measures all of your samples, from your juice, wort, or mash to the fermentation process and blending procedure to your bottled product. You can swiftly react to all undesired deviations as soon as they occur. With this lab-grade analyzer under your own roof you can always be sure that your bottles contain what their labels say, in order to keep your customer promise.Alcohol Measuring Range-Beer: 0.5 to 15 %v/v-Wine: 8 to 20 %v/v-Cider: 2 to 10 %v/v-Spirits & Liqueurs: 10 to 41 %v/v-Sake: 5 to 20 %v/vAlcohol Accuracy0.2 %v/v for beer, wine, cider, sake, and spirits with a total extract of <100 g/L0.4 %v/v for liqueurs with a total extract between 100 g/L and 450 g/L
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.