Learn More
Across International Ai -80°C 5L Capacity 316L SST Cold Trap w/ KF25 Ports ETL
SDP
Supplier:  Across International T80XUL
Encompass_Preferred
Request Bulk Order
Introducing the Ai 2nd Gen -80C (-112F) T80 cold traps with a large corrosion-resistant 316L SST 5 liter trap, 316L trapping layers/vacuum connections, and PTFE encapsulated silicone sealing gasket. Upward KF25 vacuum connections provide you safer and easier vacuum tubing connection and removal. No chilling fluid required, just plug and play.
Catalog No. 50-225-1314