Learn More
Across International Ai 50L Non-Jacketed Glass Reactor with 200°C Heating Jacket
SDP
Supplier:  Across International R50H
Encompass_Preferred
Request Bulk Order
Ai R-series single/dual jacketed glass reactors are designed for synthetic reaction of different types of materials in a controlled temperature and vacuum environment. Each reactor features adjustable stirring speeds and a constant pressure feeding funnel that allows you to add material to the vessel at a uniform and controlled speed. The built-in condenser makes it possible to recover some materials, also at a controlled rate of speed.
Catalog No. 50-225-1274