Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 617
An extremely fast filtering grade that has a larger particle retention than Grade 631. Grade 617 is a thicker filter paper with higher loading capacity used for rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions.
$36.30 - $197.10
Specifications
Particle Retention | 35 μm |
---|---|
Flow Rate | Fast |
For Use With (Application) | Rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions; construction product testing |
Grade | 617 |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
09-790-66A
|
Ahlstrom-Munksjo
61700900 |
9 cm |
Pack of 50 for $36.30
|
|
|||||
09-790-66C
|
Ahlstrom-Munksjo
61701250 |
12.5 cm |
Pack of 50 for $49.00
|
![]() |
|||||
09-790-66B
|
Ahlstrom-Munksjo
61701100 |
11 cm |
Pack of 50 for $43.05
|
|
|||||
09-790-66H
|
Ahlstrom-Munksjo
61703300 |
33 cm |
Pack of 50 for $197.10
|
|
|||||
09-790-66F
|
Ahlstrom-Munksjo
61702050 |
20.5 cm |
Pack of 50 for $85.10
|
![]() |
|||||
09-790-66E
|
Ahlstrom-Munksjo
61701850 |
18 cm |
Pack of 50 for $85.10
|
|
|||||
09-790-66D
|
Ahlstrom-Munksjo
61701500 |
15 cm |
Pack of 50 for $54.20
|
![]() |
|||||
09-790-66G
|
Ahlstrom-Munksjo
61702500 |
25 cm |
Pack of 50 for $101.70
|
![]() |
|||||
Specifications
35 μm | |
Fast | |
Rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions; construction product testing | |
617 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title