Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acetic Acid Solution
SCI_ED

Catalog Number S25840A
Click to view available options
Quantity:
1 L
Packaging:
0.1M
1.0M
6M
64-19-7, 7732-18-5
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O
64-19-7, 7732-18-5
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O

This product is for educational use - no C of A is available.

Quantity 1 L
Packaging 1.0M
Physical Form Liquid
Chemical Name or Material Acetic Acid
DOTInformation DOT Class 8, III, : Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit