Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ accumet™ Portable AP110 pH Meter
For accurate measurements anytime, anywhere.
$837.00 - $1034.00
Specifications
For Use With (Application) | pH reading |
---|---|
Material | Rugged waterproof IP67 housing |
Catalog Number | Includes | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Includes | Type | Price | Quantity & Availability | |||||
S04971
|
Meter, 9V battery, and manual | AP110 pH Meter |
Each for $837.00
|
|
|||||
S04974
|
Meter, pH/ATC electrode, refill solution, hard carrying case, sample bottles, pH calibration packets, 9V battery, and manual | AP110 pH Meter Kit |
Each for $1,034.00
|
|
|||||
Description
- Large LCD with backlighting and plain-language text
- Rugged waterproof IP67 housing
- 200 data point internal memory
- Meter Only (S04971) includes meter, 9V battery, and manual
- Meter Kit (S04974) includes meter, pH/ATC electrode, refill solution, hard carrying case, sample bottles, pH calibration packets, 9V battery, and manual
pH Mode
- Range: -2.00 to +20.00
- Resolution: 0.1/0.01
- Accuracy: ±0.01
- Calibration: Up to five or six points from threesets: (USA: 2.00, 4.00, 7.00, 10.00 12.00; NIST: 1.68, 4.01, 6.86, 9.18, 12.45; EURO: 1.00, 3.00, 6.00, 8.00, 10.00, 13.00)
Warranty and Services
Three year meter warranty, one year electrode warranty
Specifications
pH reading | |
Rugged waterproof IP67 housing |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title