Learn More
Anton Paar Digital Compact Refractometer – Abbemat 3000
Supplier:  Anton Paar 173451
Encompass_Preferred
Request Bulk Order
For measuring the concentration and refractive index of liquids and pastes, the Abbemat 3000 refractometers embody reliability, precision, and convenience. They require minimal cleaning and no maintenance. The premium technology and compactness of the Abbemat 3000 with its automated temperature-correction transform time-consuming measurements into highly efficient quality control. For routine sugar and beverage analysis- Available sugar scales, e.g. Brix, saccharose, HFCS scales- Results within 4 seconds with temperature-corrected measurementsMeasuring range Refractive index scale (RI)- Range nD 1.30 to 1.66 - Resolution nD ± 0.0001- Accuracy nD ± 0.0001Measuring range Brix scale- Range 0 to 100 %- Resolution 0.01 %- Accuracy 0.05 %Temperature- Temperature correction- Accuracy ± 0.05 °C
Catalog No. NC1818325