Learn More
Xylem Inc 50ML DOSING UNIT
Supplier: Xylem Inc 285223767
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
50 ml dosing unit complete, including piston, piston rod and UV protection coat for TITRONIC universal and TITRONIC 300.