Learn More
Chemglass Life Sciences 500ml Glassblowers Heavy Wall Filter Flask Blank
Supplier: Chemglass Life Sciences CG62302
500ml Glassblowers Heavy Wall Filter Flask Blank. Extra heavy, unmarked blanks for fabrication of filter apparatus. Top has a tooled finish to accept a rubber stopper or neoprene filter adapter.