Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Scott Fire & Safety™ Industrial SCBA - ISCBA
Self-contained breathing apparatus for industrial applications.
$3900.00
Specifications
Product Type | Industrial SCBA |
---|---|
Harness Style | Nylon polymer buckle |
Regulator Type | EZ-Flo Standard |
Catalog Number | Mfr. No. | Facepiece Type | Pressure | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Facepiece Type | Pressure | Price | Quantity & Availability | ||||
19-122-020
|
3M™ Scott Fire & Safety™
7012623394 |
None | 2216 psi | N/A | N/A | ||||
Please contact your local Fisher Scientific sales representative for pricing | |||||||||
19-122-023
|
3M™ Scott Fire & Safety™
IND114043104101 |
AV-2000 (CS) Poly | 4500 psi |
Each for $3,900.00
|
|
||||
19-122-021
|
3M™ Scott Fire & Safety™
IND112013104101 |
AV-2000 (CS) Poly | 2216 psi | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-122-022
|
3M™ Scott Fire & Safety™
IND112013103101 |
AV-3000 SureSeal, Dupont™ Kevlar™ aramid fibers, medium | 2216 psi | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||
Description
- E-Z Flo mask-mounted pressure demand regulator
- Features Top Down Convertibility concept, which allows increase in versatility and compatibility with Scott product line
- Sturdy, zinc-plated carbon steel wire-formed back frame
- Durable nylon harness system
- Two-piece polymer waist fastener
- Alligator-style shoulder clips
- Audible bell alarm is mounted on first-stage pressure reducer
- Over-the shoulder luminescent pressure gauge
- Double-locking latch and durable fabric band allows quick and easy cylinder change
Warranty and Services
Five years
Specifications
Industrial SCBA | |
Nylon polymer buckle | |
EZ-Flo Standard |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title