Learn More
Electron Microscopy Sciences 3-Well Porcelain Micro Spot Plate
Supplier:  Electron Microscopy Sciences 7156101
Encompass_Preferred
Poreclain Micro Spot Plate is ideal for microchemical applications. The plate has three concave depressions. Cavities measure 7/8" O.D. x 1/4" Deep (22 x 7 mm). Plate overall measures 3 3/8"(L) x1 3/8"(W) (85 x 34 mm)
Catalog No. 50-243-44