Learn More
Electron Microscopy Sciences 3mm Circlips Sample Holders
Supplier: Electron Microscopy Sciences EMS015
Circlips are for all EMS TEM sample holders and specimen rods. They are made from heat treated Beryllium copper. The Circlip extraction tool is for removing our Circlips from all TEM sample holders.