Learn More
Ace Glass, Inc. 3L Instatherm filter reactor body with 150mm ID grooved top flange, #80 bottom thread, 150mm IDx185mm depth, with CAPFE O-ring and power cord, 120v/9amp max
Supplier: Ace Glass, Inc. 638835
3L Instatherm coated filter reactor body with a 150mm I.D. grooved top flange for connection to a reaction head with the use of a quick release clamp, and a #80 Ace-Thred bottom thread for use with a bottom adapter with a filter support and drain valve. Instatherm provides safer, rapid heating, and can be operated at 115v with ACE Proportional Temperature Controllers, or with a Powerstat. Reactor body supplied with a CAPFE o-ring and a power cord. Dimensions: 150mm I.D. x 185mm D.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.