Learn More
Quadrex Corporation 30M. X 0.32MM ID X 3.0UM
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Quadrex Corporation 00762430W3.0F
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Fused silica GC capillary column 007-624 phase (USP G43), 30M. X 0.32MM ID X 3.0UM