Learn More
American Biotech Supply 30 Cu. Ft. General Purpose Hazardous Location Refrigerator
Supplier:  American Biotech Supply ABTERS30
Encompass_Preferred
Request Bulk Order
Standard Hazardous Material Storage (Explosion Proof) Refrigerators Meet NFPA and OSHA guidelines for Hazardous Locations, Class 1, Division II, Group C & D.
Catalog No. NC1779813