Learn More
Quadrex Corporation 25 METER X 0.32 MM X 2.0 MICRO
SDP
Supplier:  Quadrex Corporation 007FFAP25W2.0F
Encompass_Preferred
25 METER X 0.32 MM X 2.0 MICRON FILM THICKNESS GC COLUMN
Catalog No. NC1622022