Learn More
Waters Corp Flowcell, 2475
Supplier:  Waters Corp 700001618
Encompass_Preferred
The Flow Cell Holder contains the flow cell. The Flow Cell is a quartz cell designed to maximize the fluorescence signal.and to minimize the amount of stray light that could affect the signal measurement.
Catalog No. 50-792-610