Learn More
Thermo Scientific Production Chemicals and Services 2-Propanol, 67-63-0, 4x4L
Supplier: Thermo Scientific Production Chemicals and Services 907905
Molecular Formula C3H8O; Trace Impurity Level: 200 to 10ppbSEMI; CMOS Grade; Molecular Weight 60.096; Synonyms: isopropanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol; Isopropyl alcohol (IPA) is used in IPA vapor drying of wafers. The vapor dryer advantages over spin-rinse dryers include: no water spots, decreased particles on the wafer, reduced chemical consumption and much reduced wafer breakage.