Learn More
Thermo Scientific RVT5105 Savant™ Refrigerated Vapor Trap, 115V 50/60Hz
Supplier:  Thermo Scientific RVT5105115
Encompass_Preferred
Request Bulk Order
RVT5105 Savant™ Refrigerated Vapor Trap, 115V 50/60Hz, Temperature -105°C. Condense and trap a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps.
Catalog No. NC1696080