Learn More
Chemglass Life Sciences 1000mL Glassblowers Flat Bottom Flask Blank
Supplier: Chemglass Life Sciences CG61905
1000mL Glassblowers Flat Bottom Flask Blank. Blanks for fabrication of single or multi-neck flat bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.